Learn More
Biomatik Corporation GABA-B Receptor Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50241
Encompass_Preferred
GABA-B Receptor Colorimetric Cell-Based ELISA Kit for detecting GABA-B Receptor in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9UBS5. Gene ID: 2550. Target Synonyms: Gamma-aminobutyric acid type B receptor subunit 1; GABA-B receptor 1; GABA-B-R1; GABA-BR1; GABABR1; Gb1; GABBR1; GPRC3A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-6956