Learn More
Biomatik Corporation HSF1 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50295
Encompass_Preferred
HSF1 Colorimetric Cell-Based ELISA Kit for detecting HSF1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q00613. Gene ID: 3297. Target Synonyms: Heat shock factor protein 1; Heat shock transcription factor 1; HSF 1; HSTF 1; HSTF1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-7003