Learn More
Biomatik Corporation IKK-α/β Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50306
Encompass_Preferred
IKK-α/β Colorimetric Cell-Based ELISA Kit for detecting IKK-α/β in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O15111; O14920. Gene ID: 1147; 3551. Target Synonyms: CHUK; Conserved helix-loop-helix ubiquitous kinase; I kappa-B kinase alpha; I-kappa-B kinase 1; IkappaB kinase; IkBKA; IKK-A; IKK-alpha; IKK1; IKKA; Inhibitor of nuclear factor kappa-B kinase alpha subunit; kinase IKK-alpha; NFKBIKA; Nuclear factor NFkappaB inhibitor kinase alpha; I-kappa-B kinase 2; I-kappa-B-kinase beta; IkBKB; IKK-B; IKK-beta; IKK2; IKKB; kinase IKK-beta; NFKBIKB; Nuclear factor NF-kappa-B inhibitor kinase beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-7013