Learn More
Biomatik Corporation Human Collectrin(TMEM27) ELISA kit
Supplier:  Biomatik Corporation EKC42678
Encompass_Preferred
Sample Type: serum, plasma,tissue homogenates, cell lysates. Species Reactivity: Homo sapiens (Human). Detection Range: 56 pg/mL-800 pg/mL. Sensitivity: 24 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9HBJ8. Target Synonyms: NX-17, NX17, 0610008J07Rik|collectrin|kidney-specific membrane protein. Restriction: For Research Use Only.
Catalog No. 50-229-5170