Learn More
Biomatik Corporation Human Tumor Necrosis Factor Receptor Superfamily Member 18 (Tnfrsf18) Elisa Kit
Supplier:  Biomatik Corporation EKC42682
Encompass_Preferred
Human Tumor Necrosis Factor Receptor Superfamily Member 18 (Tnfrsf18) Elisa Kit for detecting Tumor Necrosis Factor Receptor Superfamily, Member 18 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9Y5U5; TNFRSF18. Target Synonyms: Activation inducible TNFR family receptor; Activation-inducible TNFR family receptor; AITR; CD357; GITR; GITR D; GITR-D; GITRD; Glucocorticoid induced TNFR related protein; glucocorticoid-induced tnf receptor ligand; glucocorticoid-induced tnf receptors ligand; Glucocorticoid-induced TNFR-related protein; TNF receptor superfamily activation inducible protein; TNFRSF 18; TNFRSF18; TNR18_HUMAN; Tumor necrosis factor receptor superfamily member 18; Tumor necrosis factor receptor superfamily member 18 precursor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5172