Learn More
Biomatik Corporation Bovine Transthyretin (Ttr) Elisa Kit
Supplier:  Biomatik Corporation EKC42700
Encompass_Preferred
Bovine Transthyretin (Ttr) Elisa Kit for detecting Transthyretin in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 312.5-5000ng/ml. Sensitivity: 312.5ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: O46375; TTR. Target Synonyms: TTR; Transthyretin; Prealbumin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5187