Learn More
Biomatik Corporation Human N-Terminal Procollagen Iv Propeptide, Pivnp Elisa Kit
Supplier:  Biomatik Corporation EKC42731
Encompass_Preferred
Human N-Terminal Procollagen Iv Propeptide, Pivnp Elisa Kit for detecting N-Terminal Procollagen IV Propeptide, PIVnp in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 2.8-180ng/ml. Sensitivity: 0.7ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: N-terminal procollagen 4 propeptide; P4NP; N-terminal procollagen IV propeptide; PIVNP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5204