Learn More
Biomatik Corporation Mouse calcitonin gene related peptide (CGRP) ELISA kit
Supplier:  Biomatik Corporation EKC42750
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q99MP3. Target Synonyms: Beta type CGRP、Calcitonin gene-related peptide II、CGRPII、Calcitonin gene-related peptide 2、CALCB. Restriction: For Research Use Only.
Catalog No. 50-229-5211