Learn More
Biomatik Corporation Mouse Anti-Cyclic Citrullinated Peptide Antibody (Anti-CCP-antibody) ELISA kit
Supplier:  Biomatik Corporation EKC42754
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Mus musculus (Mouse). Detection Range: 6.25 U/mL-400 U/mL. Sensitivity: 1.56 U/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-5213