Learn More
Biomatik Corporation Human Lysophosphatidic Acid (Lpa) Elisa Kit
Supplier:  Biomatik Corporation EKC42758
Encompass_Preferred
Human Lysophosphatidic Acid (Lpa) Elisa Kit for detecting Lysophosphatidic Acid (Lpa) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 3.9-250ng/ml. Sensitivity: 3.9ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5215