Learn More
Biomatik Corporation Fish free tri-iodothyronine (FT3) ELISA kit
Supplier:  Biomatik Corporation EKC42765
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Fish. Detection Range: 2 pmol/L-32 pmol/L. Sensitivity: 0.38 pmol/L. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-5221