Learn More
Biomatik Corporation Mouse Tumor necrosis factor receptor superfamily member 13B(TNFRSF13B) ELISA kit
Supplier:  Biomatik Corporation EKC42819
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 78 pg/ml - 5000 pg/ml. Sensitivity: 19.5 pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Not Available. Assay Time: 1-5 hours. UniProt ID: Q9ET35. Target Synonyms: CD267, CVID, FLJ39942, MGC133214, MGC39952, TACI, TNFRSF14B, transmembrane activator and CAML interactor|tumor necrosis factor receptor 13B. Restriction: For Research Use Only.
Catalog No. 50-229-5235