Learn More
Biomatik Corporation Human VGF (Neurosecretory protein VGF) ELISA Kit
Supplier:  Biomatik Corporation EKF57224
Encompass_Preferred
Human VGF (Neurosecretory protein VGF) ELISA Kit for detecting VGF in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O15240. Target Synonyms: VGF; Neurosecretory protein VGF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5260