Learn More
Biomatik Corporation Human VAV3 (Guanine nucleotide exchange factor VAV3) ELISA Kit
Supplier:  Biomatik Corporation EKF57230
Encompass_Preferred
Human VAV3 (Guanine nucleotide exchange factor VAV3) ELISA Kit for detecting VAV3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9UKW4. Target Synonyms: VAV3; Guanine nucleotide exchange factor VAV3; VAV-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5262