Learn More
Biomatik Corporation Human S1PR3 (Sphingosine 1-phosphate receptor 3) ELISA Kit
Supplier:  Biomatik Corporation EKF57243
Encompass_Preferred
Human S1PR3 (Sphingosine 1-phosphate receptor 3) ELISA Kit for detecting S1PR3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q99500. Target Synonyms: S1PR3; EDG3; LPB3; S1P3; EDG-3; Endothelial differentiation G-protein coupled receptor 3; S1P receptor 3; S1P receptor EDG3; S1P receptor Edg-3; sphingosine 1-phosphate receptor 3; Sphingosine 1-phosphate receptor Edg-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5268