Learn More
Biomatik Corporation Human ALDH1A1 (Retinal dehydrogenase 1) ELISA Kit
Supplier:  Biomatik Corporation EKF57277
Encompass_Preferred
Human ALDH1A1 (Retinal dehydrogenase 1) ELISA Kit for detecting ALDH1A1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100miu/ml. Sensitivity: 0.938miu/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P00352. Target Synonyms: RALDH1; Retinal dehydrogenase 1; ALDH1A1; ALDH-E1; PUMB1; acetaldehyde dehydrogenase 1; Aldehyde dehydrogenase family 1 member A1; ALDH class 1; ALDH1; ALDH11; ALHDII; retinaldehyde dehydrogenase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5286