Learn More
Biomatik Corporation Human NTs (Neurotensin) ELISA Kit
Supplier:  Biomatik Corporation EKF57326
Encompass_Preferred
Human NTs (Neurotensin) ELISA Kit for detecting NTs in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100pg/ml. Sensitivity: 0.938pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: P30990. Target Synonyms: NT; Neurotensin; NTS; neuromedin N; NN; NT; ; NT; N; NTS1; NMN-125. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5294