Learn More
Biomatik Corporation Human MCP/CD46 (Membrane Cofactor Protein) ELISA Kit
Supplier:  Biomatik Corporation EKF57337
Encompass_Preferred
Human MCP/CD46 (Membrane Cofactor Protein) ELISA Kit for detecting MCP; CD46 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P15529. Target Synonyms: MCP; CD46; Membrane Cofactor Protein; AHUS2; MIC10; CD46 molecule; complement regulatory protein; TLXcomplement regulatory protein; trophoblast leucocyte common antigen; trophoblast-lymphocyte cross-reactive antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5299