Learn More
Biomatik Corporation Human TTPA (Alpha-tocopherol transfer protein) ELISA Kit
Supplier:  Biomatik Corporation EKF57341
Encompass_Preferred
Human TTPA (Alpha-tocopherol transfer protein) ELISA Kit for detecting TTPA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P49638. Target Synonyms: TTPA; Alpha-tocopherol transfer protein; Alpha-TTP; TPP1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5301