Learn More
Biomatik Corporation Human CST9 (Cystatin-9) ELISA Kit
Supplier:  Biomatik Corporation EKF57349
Encompass_Preferred
Human CST9 (Cystatin-9) ELISA Kit for detecting CST9 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q5W186. Target Synonyms: CST9; Cystatin-9; Cystatin-like molecule; CLM. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5304