Learn More
Biomatik Corporation Human DMBT1 (Deleted in malignant brain tumors 1 protein) ELISA Kit
Catalog No. 502295314
Encompass_Preferred
Human DMBT1 (Deleted in malignant brain tumors 1 protein) ELISA Kit for detecting DMBT1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9UGM3. Target Synonyms: DMBT1; Deleted in malignant brain tumors 1 protein; Surfactant pulmonary-associated D-binding protein; Salivary agglutinin; SAG; Glycoprotein 340; Gp-340; Hensin; GP340; muclin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-5314 Supplier Biomatik Corporation Supplier No. EKF57375
Human DMBT1 (Deleted in malignant brain tumors 1 protein) ELISA Kit for detecting DMBT1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9UGM3. Target Synonyms: DMBT1; Deleted in malignant brain tumors 1 protein; Surfactant pulmonary-associated D-binding protein; Salivary agglutinin; SAG; Glycoprotein 340; Gp-340; Hensin; GP340; muclin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.