Learn More
Biomatik Corporation Human PYY2 (Putative peptide YY-2) ELISA Kit
Supplier:  Biomatik Corporation EKF57383
Encompass_Preferred
Human PYY2 (Putative peptide YY-2) ELISA Kit for detecting PYY2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9NRI6. Target Synonyms: PYY2; Putative peptide YY-2; Putative peptide YY2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5320