Learn More
Biomatik Corporation Human PLA2G4A (Cytosolic phospholipase A2) ELISA Kit
Supplier:  Biomatik Corporation EKF57387
Encompass_Preferred
Human PLA2G4A (Cytosolic phospholipase A2) ELISA Kit for detecting PLA2G4A in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P47712. Target Synonyms: cPLA2; PLA2G4A; Cytosolic phospholipase A2; cPLA2-alpha; calcium-dependent phospholipid-binding protein; lysophospholipase; phosphatidylcholine 2-acylhydrolase; Phospholipase A2 group IVA; phospholipase A2; group IVA; cytosolic; calcium-dependent. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5322