Learn More
Biomatik Corporation Human TNNT1 (Troponin T, slow skeletal muscle) ELISA Kit
Supplier:  Biomatik Corporation EKF57403
Encompass_Preferred
Human TNNT1 (Troponin T, slow skeletal muscle) ELISA Kit for detecting TNNT1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P13805. Target Synonyms: TNNT1; Troponin T Type 1; Slow Skeletal; TNT; TnTs; sTnT; Slow skeletal muscle troponin T; Troponin T; slow skeletal muscle. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5330