Learn More
Biomatik Corporation Human CDK2 (Cyclin-dependent kinase 2) ELISA Kit
Supplier:  Biomatik Corporation EKF57415
Encompass_Preferred
Human CDK2 (Cyclin-dependent kinase 2) ELISA Kit for detecting CDK2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P24941. Target Synonyms: CDK2; Cyclin-dependent kinase 2; Cell division protein kinase 2; p33 protein kinase; CDKN2; cdc2-related protein kinase; Cell division kinase 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5336