Learn More
Biomatik Corporation Human PROS1 (Vitamin K-dependent protein S) ELISA Kit
Supplier:  Biomatik Corporation EKF57420
Encompass_Preferred
Human PROS1 (Vitamin K-dependent protein S) ELISA Kit for detecting PROS1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400ng/ml. Sensitivity: 3.75ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P07225. Target Synonyms: PROS1; Vitamin K-dependent protein S; PSA; PROS; PS21; PS22; PS23; PS24; PS25; THPH5; THPH6; protein S; alpha; protein Sa; vitamin K-dependent plasma protein S; vitamin K-dependent protein S. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5338