Learn More
Biomatik Corporation Human MGMT (Methylated-DNA--protein-cysteine methyltransferase) ELISA Kit
Supplier:  Biomatik Corporation EKF57429
Encompass_Preferred
Human MGMT (Methylated-DNA--protein-cysteine methyltransferase) ELISA Kit for detecting MGMT in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P16455. Target Synonyms: MGMT; Methylated-DNA--protein-cysteine methyltransferase; 6-O-methylguanine-DNA methyltransferase; MGMT; O-6-methylguanine-DNA-alkyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5344