Learn More
Biomatik Corporation Human NR1H4 (Bile acid receptor) ELISA Kit
Supplier:  Biomatik Corporation EKF57434
Encompass_Preferred
Human NR1H4 (Bile acid receptor) ELISA Kit for detecting NR1H4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q96RI1. Target Synonyms: NR1H4; Bile acid receptor; Retinoid X receptor-interacting protein 14; RXR-interacting protein 14; Nuclear receptor subfamily 1 group H member 4; Farnesoid X-activated receptor; Farnesol receptor; HRR-1; BAR; FXR; HRR1; RIP14. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5347