Learn More
Biomatik Corporation Human NEFH (Neurofilament heavy polypeptide) ELISA Kit
Supplier:  Biomatik Corporation EKF57477
Encompass_Preferred
Human NEFH (Neurofilament heavy polypeptide) ELISA Kit for detecting NEFH in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P12036. Target Synonyms: Human NEFH; NEFH neurofilament; heavy polypeptide; neurofilament; heavy polypeptid; NFH; 200 kDa neurofilament protein; Neurofilament triplet H protein; NF-H. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5366