Learn More
Biomatik Corporation Human SEMA4D (Semaphorin-4D) ELISA Kit
Supplier:  Biomatik Corporation EKF57480
Encompass_Preferred
Human SEMA4D (Semaphorin-4D) ELISA Kit for detecting SEMA4D in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8000pg/ml. Sensitivity: 75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q92854. Target Synonyms: SEMA4D; GR3; A8; BB18; CD100; COLL-4; M-sema-G; SEMA4D; SEMAJ; A8; CD100 antigen; CD100M-sema-G; coll-4; immunoglobulin domain; Ig; transmembrane domain; TM and shortcytoplasmic domain; 4D; sema domain; immunoglobulin domain; Ig; transmembrane domain; TM and shortcytoplasmic domain; ; semaphorin 4D; semaphorin-4D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5369