Learn More
Biomatik Corporation Human RUNX2 (Runt-related transcription factor 2) ELISA Kit
Supplier:  Biomatik Corporation EKF57502
Encompass_Preferred
Human RUNX2 (Runt-related transcription factor 2) ELISA Kit for detecting RUNX2 in cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q13950. Target Synonyms: RUNX2; CBFA1; Acute myeloid leukemia 3 protein; AML3CCD; Core-binding factor subunit alpha-1; PEA2-alpha A; PEBP2A; PEBP2A1CCD1; PEBP2aA1; PEBP2-alpha A; runt domain; alpha subunit 1; runt-related transcription factor 2; SL3; AKV core-binding factor alpha A subunit; SL3-3 enhancer factor 1 alpha A subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5377