Learn More
Biomatik Corporation Human PEPD (Xaa-Pro dipeptidase) ELISA Kit
Supplier:  Biomatik Corporation EKF57505
Encompass_Preferred
Human PEPD (Xaa-Pro dipeptidase) ELISA Kit for detecting PEPD in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.125-200ng/ml. Sensitivity: 1.875ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P12955. Target Synonyms: PEPD; Proline dipeptidase; Prolidase; Imidodipeptidase; Peptidase D; PRD. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5379