Learn More
Biomatik Corporation Human TNFAIP3 (Tumor necrosis factor alpha-induced protein 3) ELISA Kit
Supplier:  Biomatik Corporation EKF57520
Encompass_Preferred
Human TNFAIP3 (Tumor necrosis factor alpha-induced protein 3) ELISA Kit for detecting TNFAIP3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P21580. Target Synonyms: TNFAIP3; OTUD7C; TNFA1P2; A20TNFA1P2; EC 3.4.19.12; EC 6.3.2.-; OTU domain-containing protein 7C; Putative DNA-binding protein A20; TNF alpha-induced protein 3; tumor necrosis factor alpha-induced protein 3; tumor necrosis factor inducible protein A20; tumor necrosis factor; alpha-induced protein 3; Zinc finger protein A20. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5388