Learn More
Biomatik Corporation Human HSPA2 (Heat shock-related 70 kDa protein 2) ELISA Kit
Supplier:  Biomatik Corporation EKF57524
Encompass_Preferred
Human HSPA2 (Heat shock-related 70 kDa protein 2) ELISA Kit for detecting HSPA2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P54652. Target Synonyms: HSPA2; HSP70-2; Heat shock 70 kDa protein 2; heat shock 70kD protein 2; heat shock 70kDa protein 2; heat shock-related 70 kDa protein 2; HSP70-2; HSP70-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5390