Learn More
Biomatik Corporation Human GLDC (Glycine Dehydrogenase) ELISA Kit
Supplier:  Biomatik Corporation EKF57526
Encompass_Preferred
Human GLDC (Glycine Dehydrogenase) ELISA Kit for detecting GLDC in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P23378. Target Synonyms: GLDC; GCSP; Glycine dehydrogenase; aminomethyl-transferring; Glycine cleavage system P protein; Glycine decarboxylase; Glycine dehydrogenase [decarboxylating]; mitochondrial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5392