Learn More
Biomatik Corporation Human anti AMPH (anti amphiphysin) ELISA Kit
Supplier:  Biomatik Corporation EKF57532
Encompass_Preferred
Human anti AMPH (anti amphiphysin) ELISA Kit for detecting anti-AMPH in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antigen. Assay Time: 3h, 30 min. Accession Number: P49418. Target Synonyms: anti-AMPH. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5394