Learn More
Biomatik Corporation Human ANNA1 (Anti-Neuronal Nuclear Antibody 1) ELISA Kit
Supplier:  Biomatik Corporation EKF57536
Encompass_Preferred
Human ANNA1 (Anti-Neuronal Nuclear Antibody 1) ELISA Kit for detecting ANNA1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antigen. Assay Time: 3h, 30 min. Target Synonyms: ANNA1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5397