Learn More
Biomatik Corporation Human CDNF (Cerebral Dopamine Neurotrophic Factor) ELISA Kit
Supplier:  Biomatik Corporation EKF57565
Encompass_Preferred
Human CDNF (Cerebral Dopamine Neurotrophic Factor) ELISA Kit for detecting CDNF in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q49AH0. Target Synonyms: CDNF; ARMETL1; arginine-rich; mutated in early stage tumors-like 1; ARMETL1; ARMET-like protein 1; cerebral dopamine neurotrophic factor; Conserved dopamine neurotrophic factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5408