Learn More
Biomatik Corporation Human F5 (Coagulation Factor V/Plasma Factor V) ELISA Kit
Supplier:  Biomatik Corporation EKF57601
Encompass_Preferred
Human F5 (Coagulation Factor V/Plasma Factor V) ELISA Kit for detecting F5 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P12259. Target Synonyms: F5; Coagulation Factor V; Plasma Factor V; Activated protein C cofactor; PCCF; Proaccelerin; labile factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5417