Learn More
Biomatik Corporation Human Flt3L (FMS Like Tyrosine Kinase 3 Ligand) ELISA Kit
Supplier:  Biomatik Corporation EKF57610
Encompass_Preferred
Human Flt3L (FMS Like Tyrosine Kinase 3 Ligand) ELISA Kit for detecting Flt3L in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P49771. Target Synonyms: Flt3L; FLT3LG; FL; Flt3 ligand; FLT3L; Flt3L; fms-related tyrosine kinase 3 ligand; SL cytokine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5421