Learn More
Biomatik Corporation Human GRIN2B (Glutamate Receptor, Ionotropic, N-Methyl-D-Aspartate 2B) ELISA Kit
Supplier:  Biomatik Corporation EKF57627
Encompass_Preferred
Human GRIN2B (Glutamate Receptor, Ionotropic, N-Methyl-D-Aspartate 2B) ELISA Kit for detecting GRIN2B in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q13224. Target Synonyms: GRIN2B; NMDA R; NR2B Subunit; NMDAR2B; NR2B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5427