Learn More
Biomatik Corporation Human HO1 (Heme Oxygenase 1) ELISA Kit
Supplier:  Biomatik Corporation EKF57637
Encompass_Preferred
Human HO1 (Heme Oxygenase 1) ELISA Kit for detecting HO1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P09601. Target Synonyms: HO1; HO-1; HMOX1; HSP32; heat shock protein; 32-kD; heme oxygenase; decycling 1; HO; HO1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5430