Learn More
Biomatik Corporation Human LZMg (Lysozyme G) ELISA Kit
Supplier:  Biomatik Corporation EKF57650
Encompass_Preferred
Human LZMg (Lysozyme G) ELISA Kit for detecting LZMG in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.25-80ng/ml. Sensitivity: 0.75ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antibody. Assay Time: 3h, 30 min. Accession Number: Q8N1E2. Target Synonyms: lysozyme G. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5432