Learn More
Biomatik Corporation Human NPYR2 (Neuropeptide Y Receptor Y2) ELISA Kit
Supplier:  Biomatik Corporation EKF57667
Encompass_Preferred
Human NPYR2 (Neuropeptide Y Receptor Y2) ELISA Kit for detecting NPYR2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P49146. Target Synonyms: NPYR2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5443