Learn More
Biomatik Corporation Human ST2 (Syntenin 2) ELISA Kit
Supplier:  Biomatik Corporation EKF57722
Encompass_Preferred
Human ST2 (Syntenin 2) ELISA Kit for detecting ST2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9H190. Target Synonyms: ST2; Syntenin 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5458