Learn More
Biomatik Corporation Human TAGLN (Transgelin) ELISA Kit
Supplier:  Biomatik Corporation EKF57727
Encompass_Preferred
Human TAGLN (Transgelin) ELISA Kit for detecting TAGLN in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q01995. Target Synonyms: TAGLN; SM22 alpha; TAGLN; WS3-10; Protein WS3-10; SM2222 kDa actin-binding protein; Smooth muscle protein 22-alpha; TAGLN1; transgelin; transgelin variant 2; WS3-10SM22-alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5460