Learn More
Biomatik Corporation Human tPA (Plasminogen Activator, Tissue) ELISA Kit
Supplier:  Biomatik Corporation EKF57734
Encompass_Preferred
Human tPA (Plasminogen Activator, Tissue) ELISA Kit for detecting tPA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P00750. Target Synonyms: tPA; Alteplase; PLAT; Reteplase; T-PA; Alteplase; alteplase; DKFZp686I03148; EC 3.4.21; EC 3.4.21.68; plasminogen activator; tissue; plasminogen activator; tissue type; PLAT; reteplase; Reteplase; tissue plasminogen activator; t-PA; tissue-type plasminogen activator; TPA; tPA; T-PA; t-PA; t-plasminogen activator. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5462