Learn More
Biomatik Corporation Rat SELP (P-selectin) ELISA Kit
Supplier:  Biomatik Corporation EKF57869
Encompass_Preferred
Rat SELP (P-selectin) ELISA Kit for detecting SELP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P98106. Target Synonyms: SELP; P-Selectin; CD62P; GMP140; GRMP; PADGEM; PSEL; CD62P antigen; antigen CD62; selectin P. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5468