Learn More
Biomatik Corporation Human TXNDC5 (Thioredoxin domain-containing protein 5) ELISA Kit
Supplier:  Biomatik Corporation EKF58003
Encompass_Preferred
Human TXNDC5 (Thioredoxin domain-containing protein 5) ELISA Kit for detecting TXNDC5 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8NBS9. Target Synonyms: EndoPDI; ER protein 46; ERP46; Hcc 2; PDIA15; Thioredoxin like protein p46; TLP46; TXNDC5; UNQ364. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5482