Learn More
Biomatik Corporation Mouse S100A9 (S100 Calcium Binding Protein A9) ELISA Kit
Supplier:  Biomatik Corporation EKF58022
Encompass_Preferred
Mouse S100A9 (S100 Calcium Binding Protein A9) ELISA Kit for detecting S100A9 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P31725. Target Synonyms: S100A9; Calgranulin B; MRP-14; CAGBMigRation inhibitory factor-related protein 14; calgranulin B; calgranulin-B; Calgranulin-B; Calprotectin L1H subunit; CFAGMRP-14; 60B8AG; CGLB; L1AG; LIAG; MAC387; MIF; MRP14Leukocyte L1 complex heavy chain; NIF; P14; p14; protein S100-A9; S100 calcium binding protein A9; S100 calcium binding protein A9; calgranulin B; S100 calcium-binding protein A9; S100 calcium-binding protein A9; calgranulin B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5490